missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorophenol 97.0+%, TCI America™

Catalog No. p-7117656
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19585G 5 g
A195825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19585G Supplier TCI America Supplier No. A19585G
Only null left

Chemical Identifiers

CAS 28443-50-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD02093863
InChI Key FZCQMIRJCGWWCL-UHFFFAOYSA-N
Synonym 4-chloro-2-hydroxyaniline, phenol, 2-amino-5-chloro, 2-amino-5-chloro-phenol, 5-chloro-2-aminophenol, pubchem1478, 5-chloro-o-aminophenol, 5-chloro-2-amino phenol, 2-hydroxy-4-chloroaniline, acmc-209h2j, 2-azanyl-5-chloranyl-phenol
PubChem CID 91591
ChEBI CHEBI:75051
IUPAC Name 2-amino-5-chlorophenol
SMILES NC1=CC=C(Cl)C=C1O

Specifications

Melting Point 155°C
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chlorophenol
EINECSNumber (3)-0906
TSCA Yes