missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzyl alcohol, 98%

Catalog No. AAH3053106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3053106 5 g
AAH3053114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3053106 Supplier Thermo Scientific Chemicals Supplier No. H3053106

CAS: 37585-25-4 | C7H8ClNO | 157.597 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37585-25-4
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00075163
InChI Key CLKBZWDZDVOIGJ-UHFFFAOYSA-N
PubChem CID 586754
IUPAC Name (2-amino-5-chlorophenyl)methanol
SMILES C1=CC(=C(C=C1Cl)CO)N

Specifications

Melting Point 107°C to 109°C
Quantity 5 g
Sensitivity Air Sensitive
Synonym 2-amino-5-chlorophenyl methanol, 2-amino-5-chlorobenzyl alcohol, 2-amino-5-chloro-phenyl-methanol, benzenemethanol, 2-amino-5-chloro, acmc-20ao4o, 2-amino-5-chlorobenzenemethanol, 2-hydroxymethyl-4-chloroaniline, benzenemethanol,2-amino-5-chloro, 2-amino-5-chlorophenyl methanol #
Formula Weight 157.6
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chlorobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only