accessibility menu, dialog, popup

UserName

ClOHNH2

CAS RN 37585-25-4

IUPAC Name: (2-amino-5-chlorophenyl)methanol
Synonyms: (2-amino-5-chlorophenyl)methanol
Molecular Weight (g/mol): 157.6
Molecular Formula: C7H8ClNO
InChi Key: CLKBZWDZDVOIGJ-UHFFFAOYSA-N
SMILES: NC1=C(CO)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results