missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzamide 98.0+%, TCI America™

Catalog No. A19705G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19705G 5 g
A19701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19705G Supplier TCI America Supplier No. A19705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5202-85-7
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.596
MDL Number MFCD00017126
InChI Key DNRVZOZGQHHDAT-UHFFFAOYSA-N
Synonym 5-chloroanthranilamide, benzamide, 2-amino-5-chloro, 2-amino-5-chloro-benzamide, 5-chloro-2-aminobenzamide, 2-azanyl-5-chloranyl-benzamide, pubchem13624, acmc-1axdu, 3-14-00-00963 beilstein handbook reference, 5mk
PubChem CID 78876
IUPAC Name 2-amino-5-chlorobenzamide
SMILES C1=CC(=C(C=C1Cl)C(=O)N)N

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 5 g
Formula Weight 170.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chlorobenzamide
RTECSNumber CU9090000
TSCA No