accessibility menu, dialog, popup

UserName

CAS RN 5202-85-7

IUPAC Name: 2-amino-5-chlorobenzamide
Synonyms: benzamide, 2-amino-5-chloro-
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: DNRVZOZGQHHDAT-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(Cl)=CC=C1N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)