missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzamide, 98+%

Catalog No. AAA1115303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1115303 1 g
AAA1115306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1115303 Supplier Thermo Scientific Chemicals Supplier No. A1115303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5202-85-7 | C7H7ClN2O | 170.596 g/mol

2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5'-chloro-2'-carboxamidobenzanilide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5′-chloro-2′-carboxamidobenzanilide.

Solubility
Soluble in acetone.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 5202-85-7
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.596
MDL Number MFCD00017126
InChI Key DNRVZOZGQHHDAT-UHFFFAOYSA-N
Synonym 5-chloroanthranilamide, benzamide, 2-amino-5-chloro, 2-amino-5-chloro-benzamide, 5-chloro-2-aminobenzamide, 2-azanyl-5-chloranyl-benzamide, pubchem13624, acmc-1axdu, 3-14-00-00963 beilstein handbook reference, 5mk
PubChem CID 78876
IUPAC Name 2-amino-5-chlorobenzamide
SMILES C1=CC(=C(C=C1Cl)C(=O)N)N

Specifications

Melting Point 169°C to 173°C
Quantity 1 g
Beilstein 743347
Solubility Information Soluble in acetone.
Formula Weight 170.6
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-chlorobenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-990-3
RTECSNumber CU9090000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only