missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzaldehyde, 97%

Catalog No. AAL1751506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1751506 5 g
AAL1751503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1751506 Supplier Thermo Scientific Chemicals Supplier No. L1751506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20028-53-9 | C7H6ClNO | 155.581 g/mol

2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 20028-53-9
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.581
MDL Number MFCD02093936
InChI Key KUIFMGITZFDQMP-UHFFFAOYSA-N
Synonym pubchem7364, acmc-209vm9, 3-chloro-6-aminobenzaldehyde, 2-amino-5-chloro-benzaldehyde, 2-amino-5-chlorobenz-aldehyde, 5-chloroanthranilaldehyde
PubChem CID 2773898
IUPAC Name 2-amino-5-chlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)C=O)N

Specifications

Melting Point 68°C to 74°C
Quantity 5 g
Beilstein 2716096
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 155.58
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chlorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only