CAS RN 20028-53-9
IUPAC Name:
2-amino-5-chlorobenzaldehyde
Synonyms:
2-amino-5-chlorobenzaldehyde
Molecular Weight (g/mol):
155.58
Molecular Formula:
C7H6ClNO
InChi Key:
KUIFMGITZFDQMP-UHFFFAOYSA-N
SMILES:
NC1=C(C=O)C=C(Cl)C=C1
1
–
1
of
1
results
1
2-Amino-5-chlorobenzaldehyde, 97%
CAS: 20028-53-9 Molecular Formula: C7H6ClNO Molecular Weight (g/mol): 155.581 MDL Number: MFCD02093936 InChI Key: KUIFMGITZFDQMP-UHFFFAOYSA-N Synonym: pubchem7364,acmc-209vm9,3-chloro-6-aminobenzaldehyde,2-amino-5-chloro-benzaldehyde,2-amino-5-chlorobenz-aldehyde,5-chloroanthranilaldehyde PubChem CID: 2773898 IUPAC Name: 2-amino-5-chlorobenzaldehyde SMILES: C1=CC(=C(C=C1Cl)C=O)N