missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromobenzotrifluoride 97.0+%, TCI America™

Catalog No. p-7117215
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10565G 5 g
A105625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10565G Supplier TCI America Supplier No. A10565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 445-02-3
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
MDL Number MFCD00064393
InChI Key PHXGKHTWEOPCEW-UHFFFAOYSA-N
Synonym 2-amino-5-bromobenzotrifluoride, 4-bromo-2-trifluoromethyl aniline, benzenamine, 4-bromo-2-trifluoromethyl, 4-bromo-2-trifluoromethyl-phenylamine, 2-amino-5-bromo trifluoromethyl benzene, 2-amino-5-bromo benzotrifluoride, 4-bromo-2-trifluoroaniline, 2-trifluoromethyl-4-bromoaniline, 4-bromo-alpha,alpha,alpha-trifluoro-o-toluidine, 4-bromo-2-trifluoromethyl-aniline
PubChem CID 67960
IUPAC Name 4-bromo-2-(trifluoromethyl)aniline
SMILES NC1=CC=C(Br)C=C1C(F)(F)F

Specifications

Boiling Point 102°C
Quantity 5 g
Formula Weight 240.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Amino-5-bromobenzotrifluoride