accessibility menu, dialog, popup

UserName

NH2BrFFF

CAS RN 445-02-3

IUPAC Name: 4-bromo-2-(trifluoromethyl)aniline
Synonyms: 4-bromo-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 240.02
Molecular Formula: C7H5BrF3N
InChi Key: PHXGKHTWEOPCEW-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results