missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%

Catalog No. p-7045174
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0968303 1 g
AAL0968306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0968303 Supplier Thermo Scientific Chemicals Supplier No. L0968303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4274-38-8 | C7H7ClF3NS | 229.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4274-38-8
Molecular Formula C7H7ClF3NS
Molecular Weight (g/mol) 229.65
MDL Number MFCD00042600
InChI Key FIAGYDIJZOWVAB-UHFFFAOYSA-N
Synonym 2-amino-4-trifluoromethyl benzenethiol hydrochloride, 3-amino-4-mercaptobenzotrifluoride hydrochloride, 2-amino-4-trifluoromethylthiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol hydrochloride, 2-amino-4-trifluoromethyl thiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol, chloride, pubchem2827, c7h6f3ns.hcl, dsstox_cid_31681, dsstox_rid_97532
PubChem CID 2734678
IUPAC Name 2-amino-4-(trifluoromethyl)benzenethiol;hydrochloride
SMILES [H+].[Cl-].NC1=CC(=CC=C1S)C(F)(F)F

Specifications

Melting Point 186°C to 190°C
Density 2.46
Quantity 1 g
UN Number UN1759
Beilstein 4273774
Formula Weight 229.65
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(trifluoromethyl)thiophenol hydrochloride
Hazard Category H302+H312+H332-H314-H317-H335-H373
Hazard Statement GHS H Statement
H314-H318-H373-H302-H312-H332-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 429-560-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only