CAS RN 4274-38-8
IUPAC Name:
hydrogen 2-amino-4-(trifluoromethyl)benzene-1-thiol chloride
Synonyms:
hydrogen 2-amino-4-(trifluoromethyl)benzenethiol chloride
Molecular Weight (g/mol):
229.65
Molecular Formula:
C7H7ClF3NS
InChi Key:
FIAGYDIJZOWVAB-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NC1=CC(=CC=C1S)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%
CAS: 4274-38-8 Molecular Formula: C7H7ClF3NS Molecular Weight (g/mol): 229.65 MDL Number: MFCD00042600 InChI Key: FIAGYDIJZOWVAB-UHFFFAOYSA-N Synonym: 2-amino-4-trifluoromethyl benzenethiol hydrochloride,3-amino-4-mercaptobenzotrifluoride hydrochloride,2-amino-4-trifluoromethylthiophenol hydrochloride,2-amino-4-trifluoromethyl benzene-1-thiol hydrochloride,2-amino-4-trifluoromethyl thiophenol hydrochloride,2-amino-4-trifluoromethyl benzene-1-thiol, chloride,pubchem2827,c7h6f3ns.hcl,dsstox_cid_31681,dsstox_rid_97532 PubChem CID: 2734678 IUPAC Name: 2-amino-4-(trifluoromethyl)benzenethiol;hydrochloride SMILES: [H+].[Cl-].NC1=CC(=CC=C1S)C(F)(F)F