missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-(trifluoromethyl)benzoic Acid 97.0+%, TCI America™
$156.59 - $622.59
Chemical Identifiers
| CAS | 402-13-1 |
|---|---|
| Molecular Formula | C8H6F3NO2 |
| Molecular Weight (g/mol) | 205.136 |
| MDL Number | MFCD00519334 |
| InChI Key | NQTLZJODEOHALT-UHFFFAOYSA-N |
| Synonym | 4-(Trifluoromethyl)anthranilic Acid |
| PubChem CID | 67869 |
| IUPAC Name | 2-amino-4-(trifluoromethyl)benzoic acid |
| SMILES | C1=CC(=C(C=C1C(F)(F)F)N)C(=O)O |
Chemical Identifiers
| 402-13-1 | |
| 205.136 | |
| NQTLZJODEOHALT-UHFFFAOYSA-N | |
| 67869 | |
| C1=CC(=C(C=C1C(F)(F)F)N)C(=O)O |
| C8H6F3NO2 | |
| MFCD00519334 | |
| 4-(Trifluoromethyl)anthranilic Acid | |
| 2-amino-4-(trifluoromethyl)benzoic acid |
Specifications
| 402-13-1 | |
| Yellow | |
| MFCD00519334 | |
| 4-(Trifluoromethyl)anthranilic Acid | |
| C1=CC(=C(C=C1C(F)(F)F)N)C(=O)O | |
| 205.136 | |
| 205.14 | |
| Crystalline Powder |
| 175°C | |
| C8H6F3NO2 | |
| 5 g | |
| NQTLZJODEOHALT-UHFFFAOYSA-N | |
| 2-amino-4-(trifluoromethyl)benzoic acid | |
| 67869 | |
| ≥97.0% (HPLC,T) | |
| 2-Amino-4-(trifluoromethyl)benzoic Acid |
Safety and Handling
TSCA : No