accessibility menu, dialog, popup

UserName

CAS RN 402-13-1

NH2OHOFFF

CAS RN 402-13-1

IUPAC Name: 2-amino-4-(trifluoromethyl)benzoic acid
Synonyms: 2-amino-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: NQTLZJODEOHALT-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1C(O)=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results