missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-tert-butylphenol 98.0+%, TCI America™

Catalog No. p-7117155
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A095725G 25 g
A0957250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A095725G Supplier TCI America Supplier No. A095725G
Only null left

Chemical Identifiers

CAS 1199-46-8
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00007698
InChI Key RPJUVNYXHUCRMG-UHFFFAOYSA-N
Synonym 2-amino-4-tert-butyl phenol, 2-amino-4-tert-butyl-phenol, 2-amino-4-t-butylphenol, unii-28021lmq3d, phenol, 2-amino-4-1,1-dimethylethyl, p-tert-butyl-o-aminophenol, acmc-209a4q, 4-tert-butyl-2-aminophenol, bidd:gt0022, 2-azanyl-4-tert-butyl-phenol
PubChem CID 70982
IUPAC Name 2-amino-4-tert-butylphenol
SMILES CC(C)(C)C1=CC=C(O)C(N)=C1

Specifications

Melting Point 162°C
Quantity 25 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-tert-butylphenol