accessibility menu, dialog, popup

UserName

CAS RN 1199-46-8

IUPAC Name: 2-amino-4-tert-butylphenol
Synonyms: 2-amino-4-tert-butylphenol
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: RPJUVNYXHUCRMG-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(O)C(N)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)