missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-tert-butylphenol, 97%

Catalog No. p-7026762
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2296406 5 g
AAB2296414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2296406 Supplier Thermo Scientific Chemicals Supplier No. B2296406
Only null left

CAS: 1199-46-8 | C10H15NO | 165.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1199-46-8
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00007698
InChI Key RPJUVNYXHUCRMG-UHFFFAOYSA-N
Synonym 2-amino-4-tert-butyl phenol, 2-amino-4-tert-butyl-phenol, 2-amino-4-t-butylphenol, unii-28021lmq3d, phenol, 2-amino-4-1,1-dimethylethyl, p-tert-butyl-o-aminophenol, acmc-209a4q, 4-tert-butyl-2-aminophenol, bidd:gt0022, 2-azanyl-4-tert-butyl-phenol
PubChem CID 70982
IUPAC Name 2-amino-4-tert-butylphenol
SMILES CC(C)(C)C1=CC=C(O)C(N)=C1

Specifications

Melting Point 162°C to 164°C
Quantity 5 g
Formula Weight 165.23
Percent Purity 97%
Chemical Name or Material 2-Amino-4-tert-butylphenol
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 214-844-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only