missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-phenylphenol 98.0+%, TCI America™
Supplier: TCI America A039725G
Specifications
| 2-Amino-4-phenylphenol | |
| 207°C | |
| MFCD00059187 | |
| 3-amino-4-hydroxybiphenyl, 3-aminobiphenyl-4-ol, 3-amino-1,1'-biphenyl-4-ol, 2-hydroxy-5-phenylaniline, 4-biphenylol, 3-amino, 1,1'-biphenyl-4-ol, 3-amino, phenol, 2-amino-4-phenyl, 2-amino-4-phenyl-phenol, 2-azanyl-4-phenyl-phenol | |
| C1=CC=C(C=C1)C2=CC(=C(C=C2)O)N | |
| 185.226 | |
| 185.23 | |
| Crystalline Powder |
| 1134-36-7 | |
| C12H11NO | |
| 25 g | |
| IGIDZGNPFWGICD-UHFFFAOYSA-N | |
| 2-amino-4-phenylphenol | |
| 14562 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 1134-36-7 | |
| 185.226 | |
| IGIDZGNPFWGICD-UHFFFAOYSA-N | |
| 14562 | |
| C1=CC=C(C=C1)C2=CC(=C(C=C2)O)N |
| C12H11NO | |
| MFCD00059187 | |
| 3-amino-4-hydroxybiphenyl, 3-aminobiphenyl-4-ol, 3-amino-1,1'-biphenyl-4-ol, 2-hydroxy-5-phenylaniline, 4-biphenylol, 3-amino, 1,1'-biphenyl-4-ol, 3-amino, phenol, 2-amino-4-phenyl, 2-amino-4-phenyl-phenol, 2-azanyl-4-phenyl-phenol | |
| 2-amino-4-phenylphenol |
Safety and Handling
EINECSNumber : (4)-0032
TSCA : Yes