accessibility menu, dialog, popup

UserName

CAS RN 1134-36-7

IUPAC Name: 3-amino-[1,1'-biphenyl]-4-ol
Synonyms: 3-amino-[1,1'-biphenyl]-4-ol
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: IGIDZGNPFWGICD-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1O)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results