missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-phenylphenol 98.0+%, TCI America™

Catalog No. A03975G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03975G 5 g
A03971G 1 g
A039725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A03975G Supplier TCI America Supplier No. A03975G

Chemical Identifiers

CAS 1134-36-7
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00059187
InChI Key IGIDZGNPFWGICD-UHFFFAOYSA-N
Synonym 3-amino-4-hydroxybiphenyl, 3-aminobiphenyl-4-ol, 3-amino-1,1'-biphenyl-4-ol, 2-hydroxy-5-phenylaniline, 4-biphenylol, 3-amino, 1,1'-biphenyl-4-ol, 3-amino, phenol, 2-amino-4-phenyl, 2-amino-4-phenyl-phenol, 2-azanyl-4-phenyl-phenol
PubChem CID 14562
IUPAC Name 2-amino-4-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC(=C(C=C2)O)N

Specifications

Melting Point 207°C
Quantity 5 g
Formula Weight 185.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-phenylphenol
EINECSNumber (4)-0032
TSCA Yes