missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-nitrophenol 98.0+%, TCI America™

Catalog No. A037825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A037825G 25 g
A0378100G 100 g
A0378500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A037825G Supplier TCI America Supplier No. A037825G

Chemical Identifiers

CAS 99-57-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00007695
InChI Key VLZVIIYRNMWPSN-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitroaniline, phenol, 2-amino-4-nitro, 4-nitro-2-aminophenol, p-nitro-o-aminophenol, 4-nitro-2-aminofenol, p-nitroaminofenol, 3-amino-4-hydroxynitrobenzene, 1-nitro-3-amino-4-hydroxybenzene, rodol 42, 2-amino-4-nitrofenol
PubChem CID 3613389
ChEBI CHEBI:82383
IUPAC Name 2-amino-4-nitrophenol
SMILES NC1=CC(=CC=C1O)[N+]([O-])=O

Specifications

Melting Point 146°C
Quantity 25 g
Formula Weight 154.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-nitrophenol
EINECSNumber (3)-0788
RTECSNumber SJ6300000
TSCA Yes