accessibility menu, dialog, popup

UserName

CAS RN 99-57-0

IUPAC Name: 2-amino-4-nitrophenol
Synonyms: 2-amino-4-nitrophenol
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: VLZVIIYRNMWPSN-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1O)[N+]([O-])=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)