missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals

Catalog No. AAA1396414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1396414 25 g
AAA1396422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1396414 Supplier Thermo Scientific Chemicals Supplier No. A1396414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-57-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00007695
InChI Key VLZVIIYRNMWPSN-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitroaniline, phenol, 2-amino-4-nitro, 4-nitro-2-aminophenol, p-nitro-o-aminophenol, 4-nitro-2-aminofenol, p-nitroaminofenol, 3-amino-4-hydroxynitrobenzene, 1-nitro-3-amino-4-hydroxybenzene, rodol 42, 2-amino-4-nitrofenol
PubChem CID 3613389
ChEBI CHEBI:82383
IUPAC Name 2-amino-4-nitrophenol
SMILES NC1=CC(=CC=C1O)[N+]([O-])=O

Specifications

Melting Point 141°C to 145°C
Quantity 25 g
Beilstein 776533
Formula Weight 154.13
Percent Purity 98%
Chemical Name or Material 2-Amino-4-nitrophenol
Hazard Category H302-H312-H315-H317-H319-H332-H335-H340
Hazard Statement GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
EINECSNumber 202-767-9
RTECSNumber SJ6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only