missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7117961
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25381G 1 g
A25385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25381G Supplier TCI America Supplier No. A25381G

Chemical Identifiers

CAS 2305-36-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00047853
InChI Key RPGKFFKUTVJVPY-UHFFFAOYSA-N
Synonym 4-methylanthranilic acid, benzoic acid, 2-amino-4-methyl, 2-amino-4-methyl-benzoic acid, 2-amino-p-toluic acid, 3-amino-4-carboxytoluene, 2-carboxy-5-methylaniline, p-toluic acid, 2-amino, rarechem al bo 0820, benzoic acid, 2-amino-4-methyl-9ci, 2-azanyl-4-methyl-benzoic acid
PubChem CID 75316
IUPAC Name 2-amino-4-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)C(=O)O)N

Specifications

Melting Point 171°C
Quantity 1 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methylbenzoic Acid