CAS RN 2305-36-4
IUPAC Name:
2-amino-4-methylbenzoic acid
Synonyms:
2-amino-4-methylbenzoic acid
Molecular Weight (g/mol):
151.17
Molecular Formula:
C8H9NO2
InChi Key:
RPGKFFKUTVJVPY-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C(O)=O)C(N)=C1
1
–
1
of
1
results
1
2-Amino-4-methylbenzoic Acid 98.0+%, TCI America™
CAS: 2305-36-4 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.165 MDL Number: MFCD00047853 InChI Key: RPGKFFKUTVJVPY-UHFFFAOYSA-N Synonym: 4-methylanthranilic acid,benzoic acid, 2-amino-4-methyl,2-amino-4-methyl-benzoic acid,2-amino-p-toluic acid,3-amino-4-carboxytoluene,2-carboxy-5-methylaniline,p-toluic acid, 2-amino,rarechem al bo 0820,benzoic acid, 2-amino-4-methyl-9ci,2-azanyl-4-methyl-benzoic acid PubChem CID: 75316 IUPAC Name: 2-amino-4-methylbenzoic acid SMILES: CC1=CC(=C(C=C1)C(=O)O)N