missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-fluorophenol 97.0+%, TCI America™

$736.30 - $3159.57

Chemical Identifiers

CAS 399-97-3
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD00077451
InChI Key ULDFRPKVIZMKJG-UHFFFAOYSA-N
Synonym 5-fluoro-2-hydroxyaniline, phenol, 2-amino-4-fluoro, 2-amino-4-fluoro-phenol, 4-fluoro-2-aminophenol, zlchem 493, pubchem2825, acmc-1ah2l, phenol,2-amino-4-fluoro, 2-amino-4-fluorophenol
PubChem CID 2735917
IUPAC Name 2-amino-4-fluorophenol
SMILES C1=CC(=C(C=C1F)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A24005G
View Documents
TCI America
A24005G
5 g
Each for $736.30
Only null left
 
A240025G
View Documents
TCI America
A240025G
25 g
Each for $3,159.57
Only null left
 
Description

Description

Specifications

Chemical Identifiers

399-97-3
127.118
ULDFRPKVIZMKJG-UHFFFAOYSA-N
2735917
C1=CC(=C(C=C1F)N)O
C6H6FNO
MFCD00077451
5-fluoro-2-hydroxyaniline, phenol, 2-amino-4-fluoro, 2-amino-4-fluoro-phenol, 4-fluoro-2-aminophenol, zlchem 493, pubchem2825, acmc-1ah2l, phenol,2-amino-4-fluoro, 2-amino-4-fluorophenol
2-amino-4-fluorophenol

Specifications

399-97-3
C6H6FNO
5 g
ULDFRPKVIZMKJG-UHFFFAOYSA-N
2-amino-4-fluorophenol
2735917
≥97.0% (T)
2-Amino-4-fluorophenol
146°C
MFCD00077451
5-fluoro-2-hydroxyaniline, phenol, 2-amino-4-fluoro, 2-amino-4-fluoro-phenol, 4-fluoro-2-aminophenol, zlchem 493, pubchem2825, acmc-1ah2l, phenol,2-amino-4-fluoro, 2-amino-4-fluorophenol
C1=CC(=C(C=C1F)N)O
127.118
127.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'