accessibility menu, dialog, popup

UserName

CAS RN 399-97-3

IUPAC Name: 2-amino-4-fluorophenol
Synonyms: 2-amino-4-fluorophenol
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: ULDFRPKVIZMKJG-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)