missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-fluorophenol 97.0+%, TCI America™

Catalog No. A24005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24005G 5 g
A240025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24005G Supplier TCI America Supplier No. A24005G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 399-97-3
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD00077451
InChI Key ULDFRPKVIZMKJG-UHFFFAOYSA-N
Synonym 5-fluoro-2-hydroxyaniline, phenol, 2-amino-4-fluoro, 2-amino-4-fluoro-phenol, 4-fluoro-2-aminophenol, zlchem 493, pubchem2825, acmc-1ah2l, phenol,2-amino-4-fluoro, 2-amino-4-fluorophenol
PubChem CID 2735917
IUPAC Name 2-amino-4-fluorophenol
SMILES C1=CC(=C(C=C1F)N)O

Specifications

Melting Point 146°C
Quantity 5 g
Formula Weight 127.12
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-fluorophenol