missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorobenzoic acid, 98%

Catalog No. AC103320250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC103320250 25 g
AC103321000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103320250 Supplier Thermo Scientific Chemicals Supplier No. 103320250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89-77-0 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-77-0
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007778
InChI Key JYYLQSCZISREGY-UHFFFAOYSA-N
Synonym 4-chloroanthranilic acid, benzoic acid, 2-amino-4-chloro, 4-chloro-2-aminobenzoic acid, anthranilic acid, 4-chloro, 4-chloroanthranil acid, 4-chloro anthranilic acid, 2-carboxy-5-chloroaniline, unii-g55k85r12h, 2-amino-4-chloro-benzoic acid, 2,4-acba
PubChem CID 66646
IUPAC Name 2-amino-4-chlorobenzoic acid
SMILES NC1=CC(Cl)=CC=C1C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 231°C to 235°C
Color Beige to Brown
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula H2NC6H3(Cl)CO2H
Quantity 25 g
Beilstein 14, 365
Solubility Information Solubility in water: slightly soluble
Formula Weight 171.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-4-chlorobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-938-5

RUO – Research Use Only