accessibility menu, dialog, popup

UserName

CAS RN 89-77-0

IUPAC Name: 2-amino-4-chlorobenzoic acid
Synonyms: 2-amino-4-chlorobenzoic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: JYYLQSCZISREGY-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC=C1C(O)=O
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results