missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chloro-6-methoxypyrimidine 95.0+%, TCI America™
Supplier: TCI America A15205G
Specifications
| 2-Amino-4-chloro-6-methoxypyrimidine | |
| 170°C | |
| C5H6ClN3O | |
| 5 g | |
| VFEYBTFCBZMBAU-UHFFFAOYSA-N | |
| 4-chloro-6-methoxypyrimidin-2-amine | |
| 79811 | |
| ≥95.0% (HPLC,T) |
| 5734-64-5 | |
| Yellow | |
| MFCD00233535 | |
| 2-amino-4-chloro-6-methoxypyrimidine, 2-pyrimidinamine, 4-chloro-6-methoxy, 2-amino-6-chloro-4-methoxypyrimidine, pyrimidine, 2-amino-4-chloro-6-methoxy, 2-amino-4-methoxy-6-chloro pyrimidine, 4-chloro-6-methoxy-2-pyrimidinamine, 2-amino-4-chloro-6-methoxylpyrimidine, 2-amino-4-chloro-6-methoxy pyrimidine, 2-amino-4-chloro-6-methoxy-pyrimidine, 4-chloro-6-methoxy-pyrimidin-2-ylamine | |
| COC1=CC(Cl)=NC(N)=N1 | |
| 159.57 | |
| 159.57 | |
| Crystalline Powder |
Chemical Identifiers
| 5734-64-5 | |
| 159.57 | |
| VFEYBTFCBZMBAU-UHFFFAOYSA-N | |
| 79811 | |
| COC1=CC(Cl)=NC(N)=N1 |
| C5H6ClN3O | |
| MFCD00233535 | |
| 2-amino-4-chloro-6-methoxypyrimidine, 2-pyrimidinamine, 4-chloro-6-methoxy, 2-amino-6-chloro-4-methoxypyrimidine, pyrimidine, 2-amino-4-chloro-6-methoxy, 2-amino-4-methoxy-6-chloro pyrimidine, 4-chloro-6-methoxy-2-pyrimidinamine, 2-amino-4-chloro-6-methoxylpyrimidine, 2-amino-4-chloro-6-methoxy pyrimidine, 2-amino-4-chloro-6-methoxy-pyrimidine, 4-chloro-6-methoxy-pyrimidin-2-ylamine | |
| 4-chloro-6-methoxypyrimidin-2-amine |
Safety and Handling
RTECSNumber : UV6260335
TSCA : Yes