accessibility menu, dialog, popup

UserName

CAS RN 5734-64-5

IUPAC Name: 4-chloro-6-methoxypyrimidin-2-amine
Synonyms: 4-chloro-6-methoxypyrimidin-2-amine
Molecular Weight (g/mol): 159.57
Molecular Formula: C5H6ClN3O
InChi Key: VFEYBTFCBZMBAU-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=NC(N)=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)