missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-methoxypyrimidine 95.0+%, TCI America™

Catalog No. p-7117493
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
A15205G 5 g
A152025G 25 g
2 options

Catalog No. A15205G

Supplier: TCI America A15205G

Only null left

Chemical Identifiers

CAS 5734-64-5
Molecular Formula C5H6ClN3O
Molecular Weight (g/mol) 159.57
MDL Number MFCD00233535
InChI Key VFEYBTFCBZMBAU-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-6-methoxypyrimidine, 2-pyrimidinamine, 4-chloro-6-methoxy, 2-amino-6-chloro-4-methoxypyrimidine, pyrimidine, 2-amino-4-chloro-6-methoxy, 2-amino-4-methoxy-6-chloro pyrimidine, 4-chloro-6-methoxy-2-pyrimidinamine, 2-amino-4-chloro-6-methoxylpyrimidine, 2-amino-4-chloro-6-methoxy pyrimidine, 2-amino-4-chloro-6-methoxy-pyrimidine, 4-chloro-6-methoxy-pyrimidin-2-ylamine
PubChem CID 79811
IUPAC Name 4-chloro-6-methoxypyrimidin-2-amine
SMILES COC1=CC(Cl)=NC(N)=N1

Specifications

Melting Point 170°C
Color Yellow
Quantity 5 g
Formula Weight 159.57
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloro-6-methoxypyrimidine
RTECSNumber UV6260335
TSCA Yes