missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-bromobenzoic acid, 97%

Catalog No. p-7037182
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55829MD 250 mg
AAH5582903 1 g
AAH5582906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55829MD Supplier Thermo Scientific Chemicals Supplier No. H55829MD

CAS: 20776-50-5 | C7H6BrNO2 | 216.034 g/mol

2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (slightly).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 20776-50-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD03618454
InChI Key BNNICQAVXPXQAH-UHFFFAOYSA-N
Synonym 4-bromoanthranilic acid, benzoic acid, 2-amino-4-bromo, 5-bromo-2-carboxyaniline, 2-amino-4-bromo-benzoic acid, 2-amino-4-brombenzoic acid, zlchem 336, pubchem13462, acmc-1cb3t, ksc207e6b, pharmabridge p-1344
PubChem CID 88691
IUPAC Name 2-amino-4-bromobenzoic acid
SMILES C1=CC(=C(C=C1Br)N)C(=O)O

Specifications

Melting Point 230°C to 234°C
Quantity 250 mg
Solubility Information Soluble in water (slightly).
Formula Weight 216.03
Percent Purity 97%
Chemical Name or Material 2-Amino-4-bromobenzoic acid
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 244-025-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only