missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4,5-dimethylbenzoic Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 208°C |
| Color | Yellow |
| Quantity | 200 mg |
| Formula Weight | 165.19 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Amino-4,5-dimethylbenzoic Acid |
Safety and Handling
| TSCA | No |