missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-(3-pyridyl)thiazole, 97%, Thermo Scientific™
$303.63 - $303.63
Chemical Identifiers
| CAS | 30235-27-9 |
|---|---|
| Molecular Formula | C8H7N3S |
| Molecular Weight (g/mol) | 177.225 |
| MDL Number | MFCD00218013 |
| InChI Key | XOHZQGAYUHOJPR-UHFFFAOYSA-N |
| Synonym | 4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamine |
| PubChem CID | 154893 |
| IUPAC Name | 4-pyridin-3-yl-1,3-thiazol-2-amine |
| SMILES | C1=CC(=CN=C1)C2=CSC(=N2)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5863004
|
Thermo Scientific Chemicals
H5863004 |
2 g |
Each for $303.63
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as a pharmaceutical intermediate.
Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
Chemical Identifiers
| 30235-27-9 | |
| 177.225 | |
| XOHZQGAYUHOJPR-UHFFFAOYSA-N | |
| 154893 | |
| C1=CC(=CN=C1)C2=CSC(=N2)N |
| C8H7N3S | |
| MFCD00218013 | |
| 4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamine | |
| 4-pyridin-3-yl-1,3-thiazol-2-amine |
Specifications
| 30235-27-9 | |
| C8H7N3S | |
| 2 g | |
| XOHZQGAYUHOJPR-UHFFFAOYSA-N | |
| 4-pyridin-3-yl-1,3-thiazol-2-amine | |
| 154893 | |
| 97% | |
| 2-Amino-4-(3-pyridyl)thiazole |
| Brown | |
| MFCD00218013 | |
| 4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamine | |
| C1=CC(=CN=C1)C2=CSC(=N2)N | |
| 177.225 | |
| 177.22 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures