missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(3-pyridyl)thiazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5863004

 View more versions of this product

Catalog No. AAH5863004

Only null left
Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
Specifications

Specifications

2-Amino-4-(3-pyridyl)thiazole
Brown
MFCD00218013
4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamine
C1=CC(=CN=C1)C2=CSC(=N2)N
177.225
177.22
Solid
30235-27-9
C8H7N3S
2 g
XOHZQGAYUHOJPR-UHFFFAOYSA-N
4-pyridin-3-yl-1,3-thiazol-2-amine
154893
97%

Chemical Identifiers

30235-27-9
177.225
XOHZQGAYUHOJPR-UHFFFAOYSA-N
154893
C1=CC(=CN=C1)C2=CSC(=N2)N
C8H7N3S
MFCD00218013
4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamine
4-pyridin-3-yl-1,3-thiazol-2-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions