missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog No. AC443180100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443180100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC443180100 Supplier Thermo Scientific Chemicals Supplier No. 443180100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 606-18-8 | Molecular Formula: C7H6N2O4 | Molecular Weight: 182.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 606-18-8
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.13
InChI Key JJPIVRWTAGQTPQ-UHFFFAOYSA-N
Synonym 2-amino-3-nitrobenzoic acid, 3-nitroanthranilic acid, benzoic acid, 2-amino-3-nitro, 2-amino-3-nitrobenzonic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitro benzoic acid, pubchem2233, amino-3-nitrobenzoic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline
PubChem CID 219633
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O

Specifications

Melting Point 204.0°C to 207.0°C
Assay Percent Range 96% min. (HPLC)
Quantity 10 g
Formula Weight 182.13
Percent Purity 97%
Chemical Name or Material 2-Amino-3-nitrobenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or

RUO – Research Use Only