accessibility menu, dialog, popup

UserName

CAS RN 606-18-8

IUPAC Name: 2-amino-3-nitrobenzoate
Synonyms: 2-amino-3-nitrobenzoate
Molecular Weight (g/mol): 181.13
Molecular Formula: C7H5N2O4
InChi Key: JJPIVRWTAGQTPQ-UHFFFAOYSA-M
SMILES: NC1=C(C=CC=C1[N+]([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results