missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chloro-1,4-naphthoquinone 98.0+%, TCI America™

Catalog No. p-7117351
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A128825G 25 g
A1288500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A128825G Supplier TCI America Supplier No. A128825G
Only null left

Chemical Identifiers

CAS 2797-51-5
Molecular Formula C10H6ClNO2
Molecular Weight (g/mol) 207.61
MDL Number MFCD00001680
InChI Key OBLNWSCLAYSJJR-UHFFFAOYSA-N
Synonym 2-amino-3-chloro-1,4-naphthoquinone, quinoclamine, quinoclamin, mogeton, 06k-quinone, acnq, o 6k-quinone, mogeton granule, mogeton g, 1,4-naphthalenedione, 2-amino-3-chloro
PubChem CID 17748
ChEBI CHEBI:81849
IUPAC Name 2-amino-3-chloro-1,4-dihydronaphthalene-1,4-dione
SMILES NC1=C(Cl)C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 200°C
Quantity 25 g
Formula Weight 207.61
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-chloro-1,4-naphthoquinone
EINECSNumber (4)-0390
RTECSNumber QL7350000
TSCA No