missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-chloro-1,4-naphthoquinone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 200°C |
| Quantity | 25 g |
| Formula Weight | 207.61 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Amino-3-chloro-1,4-naphthoquinone |
Safety and Handling
| EINECSNumber | (4)-0390 |
| RTECSNumber | QL7350000 |
| TSCA | No |