missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,6-difluorobenzonitrile, 97%

Catalog No. AAH59362MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59362MD 250 mg
AAH5936203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59362MD Supplier Thermo Scientific Chemicals Supplier No. H59362MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 190011-81-5 | C7H4F2N2 | 154.12 g/mol

2-Amino-3,6-difluorobenzonitrile is used in cosmetic and pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,6-difluorobenzonitrile is used in cosmetic and pharmaceutical intermediates

Solubility
Miscible with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 190011-81-5
Molecular Formula C7H4F2N2
Molecular Weight (g/mol) 154.12
MDL Number MFCD13178244
InChI Key ZPQFHGOBMGGBEP-UHFFFAOYSA-N
Synonym 2-amino-3,6-difluoro-benzonitrile, 2-cyano-3,6-difluoroaniline, 2-amino-3,6-difluorobenzonitrile, benzonitrile, 2-amino-3,6-difluoro-9ci
PubChem CID 45090591
IUPAC Name 2-amino-3,6-difluorobenzonitrile
SMILES NC1=C(F)C=CC(F)=C1C#N

Specifications

Quantity 250 mg
UN Number UN3439
Solubility Information Miscible with water.
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2-Amino-3,6-difluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only