CAS RN 190011-81-5
IUPAC Name:
2-amino-3,6-difluorobenzonitrile
Synonyms:
2-amino-3,6-difluorobenzonitrile
Molecular Weight (g/mol):
154.12
Molecular Formula:
C7H4F2N2
InChi Key:
ZPQFHGOBMGGBEP-UHFFFAOYSA-N
SMILES:
NC1=C(F)C=CC(F)=C1C#N
1
–
1
of
1
results
1
Promotions Available
2-Amino-3,6-difluorobenzonitrile, 97%
2-Amino-3,6-difluorobenzonitrile, 97%
CAS: 190011-81-5 Molecular Formula: C7H4F2N2 Molecular Weight (g/mol): 154.12 MDL Number: MFCD13178244 InChI Key: ZPQFHGOBMGGBEP-UHFFFAOYSA-N Synonym: 2-amino-3,6-difluoro-benzonitrile,2-cyano-3,6-difluoroaniline,2-amino-3,6-difluorobenzonitrile,benzonitrile, 2-amino-3,6-difluoro-9ci PubChem CID: 45090591 IUPAC Name: 2-amino-3,6-difluorobenzonitrile SMILES: NC1=C(F)C=CC(F)=C1C#N