missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,4-dimethylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7128108
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41205G 5 g
D412025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41205G Supplier TCI America Supplier No. D41205G
Only null left

Chemical Identifiers

CAS 50419-58-4
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00130041
InChI Key MUOBMUYSNYMSDM-UHFFFAOYSA-N
Synonym 2-amino-3,4-dimethyl-benzoic acid, 3,4-dimethylanthranilic acid, benzoic acid, 2-amino-3,4-dimethyl, pubchem4683, acmc-1aomr, ksc272a3h, 2-amino-3,4-dimethyl benzoic acid
PubChem CID 282450
IUPAC Name 2-amino-3,4-dimethylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(N)=C1C

Specifications

Melting Point 181°C
Color Yellow
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3,4-dimethylbenzoic Acid