accessibility menu, dialog, popup

UserName

CH3OHOH2NH3C

CAS RN 50419-58-4

IUPAC Name: 2-amino-3,4-dimethylbenzoic acid
Synonyms: 2-amino-3,4-dimethylbenzoic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: MUOBMUYSNYMSDM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(O)=O)C(N)=C1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results