Learn More
2-Amino-3,4,5-trimethoxybenzoic acid, 97%
CAS: 61948-85-4 | C10H13NO5 | 227.22 g/mol
$139.29 - $139.29
Chemical Identifiers
| CAS | 61948-85-4 |
|---|---|
| Molecular Formula | C10H13NO5 |
| MDL Number | MFCD00051630 |
| InChI Key | JSHSRQCOCMIIPA-UHFFFAOYSA-N |
| PubChem CID | 96602 |
| IUPAC Name | 2-amino-3,4,5-trimethoxybenzoic acid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1404206
|
Thermo Scientific Chemicals
A1404206 |
5 g |
Each for $139.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 61948-85-4 | |
| MFCD00051630 | |
| 96602 |
| C10H13NO5 | |
| JSHSRQCOCMIIPA-UHFFFAOYSA-N | |
| 2-amino-3,4,5-trimethoxybenzoic acid |
Specifications
| 61948-85-4 | |
| C10H13NO5 | |
| 5 g | |
| 3,4,5-trimethoxyanthranilic acid, benzoic acid,2-amino-3,4,5-trimethoxy, benzoic acid, 2-amino-3,4,5-trimethoxy, maybridge1_001555, acmc-1b683, 2-amino-3,4,5-trimethoxybenzoicacid, 2-amino-3,4,5-trimethoxybenzoic acid | |
| COC1=C(C(=C(C(=C1)C(=O)O)N)OC)OC | |
| 227.216 | |
| 227.22 | |
| 2-Amino-3,4,5-trimethoxybenzoic acid |
| 137°C to 141°C | |
| MFCD00051630 | |
| 3353856 | |
| JSHSRQCOCMIIPA-UHFFFAOYSA-N | |
| 2-amino-3,4,5-trimethoxybenzoic acid | |
| 96602 | |
| 0.97 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
H315-H317-H319
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 263-344-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only