missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,4,5-trimethoxybenzoic acid, 97%

Catalog No. aaa1404206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1404206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1404206 Supplier Thermo Scientific Chemicals Supplier No. A1404206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61948-85-4 | C10H13NO5 | 227.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 61948-85-4
Molecular Formula C10H13NO5
MDL Number MFCD00051630
InChI Key JSHSRQCOCMIIPA-UHFFFAOYSA-N
PubChem CID 96602
IUPAC Name 2-amino-3,4,5-trimethoxybenzoic acid

Specifications

Melting Point 137°C to 141°C
Quantity 5 g
Beilstein 3353856
Synonym 3,4,5-trimethoxyanthranilic acid, benzoic acid,2-amino-3,4,5-trimethoxy, benzoic acid, 2-amino-3,4,5-trimethoxy, maybridge1_001555, acmc-1b683, 2-amino-3,4,5-trimethoxybenzoicacid, 2-amino-3,4,5-trimethoxybenzoic acid
SMILES COC1=C(C(=C(C(=C1)C(=O)O)N)OC)OC
Molecular Weight (g/mol) 227.216
Formula Weight 227.22
Percent Purity 0.97
Chemical Name or Material 2-Amino-3,4,5-trimethoxybenzoic acid
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 263-344-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only