missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Amino-2'-deoxyguanosine, 98%

CAS: 60966-26-9 | C10H14N6O4 | 282.26 g/mol

$323.82 - $1,152.51

Chemical Identifiers

CAS 60966-26-9
Molecular Formula C10H14N6O4
Molecular Weight (g/mol) 282.26
MDL Number MFCD01723954
InChI Key ROPTVRLUGSPXNH-SJLGBTOWNA-N
PubChem CID 173775
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ64650MD
View Documents Promotion Details
Thermo Scientific Chemicals
J64650MD
250 mg
Each for $323.82
Only null left
 
AAJ6465003
View Documents Promotion Details
Thermo Scientific Chemicals
J6465003
1 g
Each for $1,152.51
Only null left
 
Description

Description

2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Chemical Identifiers

60966-26-9
282.26
ROPTVRLUGSPXNH-SJLGBTOWNA-N
2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C10H14N6O4
MFCD01723954
173775
N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O

Specifications

60966-26-9
C10H14N6O4
2'-amino-2'-deoxyguanosine, guanosine, 2'-amino-2'-deoxy, 2'-deoxy-2'-aminoguanosine, 2ag, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, guanosine,2'-amino-2'-deoxy, 2'-amino-2'-deoxyguanosine,2'-nh2-dg, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one
N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
282.26
282.26
Powder
White
MFCD01723954
ROPTVRLUGSPXNH-SJLGBTOWNA-N
2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
173775
98%
2'-Amino-2'-deoxyguanosine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

RTECSNumber : MF8752000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only