Learn More
Thermo Scientific Chemicals 2'-Amino-2'-deoxyguanosine, 98%
CAS: 60966-26-9 | C10H14N6O4 | 282.26 g/mol
Supplier: Thermo Scientific Chemicals J6465003
| Quantity | 1 g |
|---|
Description
2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
| 60966-26-9 | |
| 282.26 | |
| ROPTVRLUGSPXNH-SJLGBTOWNA-N | |
| 2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
| C10H14N6O4 | |
| MFCD01723954 | |
| 173775 | |
| N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O |
Specifications
| 2'-Amino-2'-deoxyguanosine | |
| White | |
| 1 g | |
| 2'-amino-2'-deoxyguanosine, guanosine, 2'-amino-2'-deoxy, 2'-deoxy-2'-aminoguanosine, 2ag, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, guanosine,2'-amino-2'-deoxy, 2'-amino-2'-deoxyguanosine,2'-nh2-dg, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one | |
| N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O | |
| 282.26 | |
| 282.26 | |
| Powder |
| 60966-26-9 | |
| C10H14N6O4 | |
| MFCD01723954 | |
| ROPTVRLUGSPXNH-SJLGBTOWNA-N | |
| 2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one | |
| 173775 | |
| 98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
RTECSNumber : MF8752000
TSCA : No
Recommended Storage : Keep cold