missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2',5-dichlorobenzophenone 98.0+%, TCI America™

Catalog No. A112810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A112810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A112810G Supplier TCI America Supplier No. A112810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2958-36-3
Molecular Formula C13H9Cl2NO
Molecular Weight (g/mol) 266.121
MDL Number MFCD00007840
InChI Key KWZYIAJRFJVQDO-UHFFFAOYSA-N
Synonym 2-amino-2',5-dichlorobenzophenone, 2-amino-5-chlorophenyl 2-chlorophenyl methanone, 4-chloro-2-2-chlorobenzoyl aniline, methanone, 2-amino-5-chlorophenyl 2-chlorophenyl, unii-g806veo3ke, 2-amino-5-chloro-2'-chlorobenzophenone, 2-amino-5-chlorophenyl-2-chlorophenyl methanone, g806veo3ke, benzophenone, 2-amino-2',5-dichloro, 2-amino-5,2'-dichlorobenzophenone
PubChem CID 18069
IUPAC Name (2-amino-5-chlorophenyl)-(2-chlorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Cl)N)Cl

Specifications

Melting Point 87°C
Color Yellow
Quantity 10 g
Formula Weight 266.12
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2′,5-dichlorobenzophenone
EINECSNumber (4)-0138
RTECSNumber DJ0200000
TSCA Yes